The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030891.4 | NP_112153.1 | 2 - 2 | 1809 | 62 |
GenBank mRNA | AK127604.1 | - | 3 - 3 | 1426 | 62 |
BC064321.1 | - | 2 - 2 | 1809 | 62 |