Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DSG2
Entrez Gene ID
1829
Gene Name
desmoglein 2
Gene Aliases
CDHF5, HDGC
Chromosome Location
Chr.18: 31498004 - 31549008 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001943.4 NP_001934.2 11 - 12 1897 68
GenBank mRNA AK308408.1 - 11 - 12 1654 68
BC099655.3 - 11 - 12 1660 68
BC099656.3 - 11 - 12 1660 68
BC099657.3 - 11 - 12 1660 68
Z26317.1 - 11 - 12 1657 68

View Full Product Details