The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001943.4 | NP_001934.2 | 4 - 5 | 628 | 116 |
GenBank mRNA | AK308408.1 | - | 4 - 5 | 385 | 116 |
BC099655.3 | - | 4 - 5 | 391 | 116 | |
BC099656.3 | - | 4 - 5 | 391 | 116 | |
BC099657.3 | - | 4 - 5 | 391 | 116 | |
BP353513.1 | - | 4 - 5 | 423 | 116 | |
DC417739.1 | - | 4 - 5 | 430 | 116 | |
Z26317.1 | - | 4 - 5 | 388 | 116 |