Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PEX16
Entrez Gene ID
9409
Gene Name
peroxisomal biogenesis factor 16
Gene Aliases
PBD8A, PBD8B
Chromosome Location
Chr.11: 45909669 - 45918123 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004813.2 NP_004804.1 7 - 8 1013 91
NM_057174.2 NP_476515.1 7 - 8 1013 91
XM_011520474.1 XP_011518776.1 7 - 8 752 91
GenBank mRNA AB016531.1 - 7 - 8 738 91
AF118240.1 - 7 - 8 945 91
BC000467.2 - 7 - 8 719 91
BC004356.1 - 7 - 8 711 91

View Full Product Details