The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007024.1 | NP_001007025.1 | 6 - 7 | 554 | 109 |
NM_001007025.1 | NP_001007026.1 | 6 - 7 | 532 | 109 | |
NM_004871.2 | NP_004862.1 | 6 - 7 | 538 | 109 | |
XM_005258070.4 | XP_005258127.1 | 6 - 7 | 779 | 109 | |
XM_005258071.4 | XP_005258128.1 | 6 - 7 | 772 | 109 | |
XM_005258072.2 | XP_005258129.1 | 6 - 7 | 566 | 109 | |
GenBank mRNA | AF073926.1 | - | 6 - 7 | 524 | 109 |
AK000606.1 | - | 6 - 7 | 538 | 109 | |
AK291381.1 | - | 6 - 7 | 538 | 109 | |
BC040471.1 | - | 6 - 7 | 519 | 109 | |
BG654545.1 | - | 6 - 7 | 511 | 109 |