The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080546.4 | NP_536856.2 | 15 - 16 | 2211 | 73 |
XM_006717028.3 | XP_006717091.1 | 15 - 16 | 2024 | 73 | |
XM_017014560.1 | XP_016870049.1 | 16 - 17 | 2189 | 73 | |
GenBank mRNA | AJ420812.1 | - | 15 - 16 | 1997 | 73 |
AK022549.1 | - | 11 - 12 | 1564 | 73 | |
BC018213.1 | - | 4 - 5 | 526 | 73 | |
BC049203.1 | - | 15 - 16 | 1964 | 73 |