Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRYM
Entrez Gene ID
1428
Gene Name
crystallin mu
Gene Aliases
DFNA40, THBP
Chromosome Location
Chr.16: 21258518 - 21303136 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001888.4 NP_001879.1 4 - 5 677 73
GenBank mRNA AK290852.1 - 2 - 3 396 73
AK310424.1 - 2 - 3 350 73
BC018061.1 - 2 - 3 395 73
BI860684.1 - 3 - 4 350 73
BX648477.1 - 4 - 5 593 73
L02950.1 - 2 - 3 356 73
U85772.1 - 2 - 3 351 73

View Full Product Details