The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016281.3 | NP_057365.3 | 19 - 20 | 2848 | 86 |
XM_005253897.1 | XP_005253954.1 | 19 - 20 | 2846 | 86 | |
XM_005253898.1 | XP_005253955.1 | 19 - 20 | 2737 | 86 | |
XM_006719445.2 | XP_006719508.1 | 19 - 20 | 2774 | 86 | |
XM_011538437.1 | XP_011536739.1 | 19 - 20 | 2792 | 86 | |
XM_011538439.1 | XP_011536741.1 | 19 - 20 | 2765 | 86 | |
XM_017019408.1 | XP_016874897.1 | 20 - 21 | 3057 | 86 | |
XM_017019409.1 | XP_016874898.1 | 20 - 21 | 3181 | 86 | |
XM_017019410.1 | XP_016874899.1 | 20 - 21 | 3177 | 86 | |
XM_017019411.1 | XP_016874900.1 | 20 - 21 | 3001 | 86 | |
XM_017019412.1 | XP_016874901.1 | 19 - 20 | 3868 | 86 | |
XM_017019413.1 | XP_016874902.1 | 19 - 20 | 3841 | 86 | |
GenBank mRNA | AF135158.1 | - | 19 - 20 | 2652 | 86 |
AF179867.1 | - | 19 - 20 | 2558 | 86 | |
AF181985.1 | - | 19 - 20 | 2649 | 86 | |
AF263311.1 | - | 17 - 18 | 2357 | 86 | |
AF328731.1 | - | 8 - 9 | 1449 | 86 | |
AK056370.1 | - | 6 - 7 | 1767 | 86 | |
AK289558.1 | - | 19 - 20 | 2792 | 86 | |
AL833460.1 | - | 15 - 16 | 2944 | 86 | |
AL833731.1 | - | 11 - 12 | 1789 | 86 | |
BC002756.2 | - | 19 - 20 | 2759 | 86 | |
BT007185.1 | - | 17 - 18 | 2357 | 86 |