The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079872.1 | NP_001073341.1 | 11 - 12 | 1954 | 95 |
NM_003588.3 | NP_003579.3 | 13 - 14 | 2121 | 95 | |
XM_005262481.1 | XP_005262538.1 | 13 - 14 | 2121 | 95 | |
XM_006724784.1 | XP_006724847.1 | 12 - 13 | 1789 | 95 | |
XM_006724785.2 | XP_006724848.1 | 12 - 13 | 1700 | 95 | |
XM_011531399.2 | XP_011529701.1 | 11 - 12 | 1196 | 95 | |
XM_011531400.2 | XP_011529702.1 | 11 - 12 | 1219 | 95 | |
XM_011531401.1 | XP_011529703.1 | 11 - 12 | 1057 | 95 | |
GenBank mRNA | AB014595.2 | - | 11 - 12 | 1288 | 95 |
AF212995.1 | - | 11 - 12 | 1694 | 95 | |
AK123688.1 | - | 11 - 12 | 1798 | 95 | |
AK299081.1 | - | 11 - 12 | 1225 | 95 | |
AK315037.1 | - | 13 - 14 | 2057 | 95 | |
AY365125.1 | - | 11 - 12 | 1630 | 95 | |
BC036216.1 | - | 13 - 14 | 2086 | 95 | |
BX537787.1 | - | 11 - 12 | 1945 | 95 | |
BX647096.1 | - | 11 - 12 | 1954 | 95 | |
CR936722.1 | - | 11 - 12 | 1954 | 95 |