The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079872.1 | NP_001073341.1 | 2 - 3 | 995 | 149 |
NM_003588.3 | NP_003579.3 | 4 - 5 | 1162 | 149 | |
XM_005262481.1 | XP_005262538.1 | 4 - 5 | 1162 | 149 | |
XM_006724784.1 | XP_006724847.1 | 3 - 4 | 830 | 149 | |
XM_006724785.2 | XP_006724848.1 | 3 - 4 | 741 | 149 | |
XM_011531399.2 | XP_011529701.1 | 2 - 3 | 237 | 149 | |
XM_011531400.2 | XP_011529702.1 | 2 - 3 | 260 | 149 | |
GenBank mRNA | AB014595.2 | - | 2 - 3 | 329 | 149 |
AF212995.1 | - | 2 - 3 | 735 | 149 | |
AK123688.1 | - | 2 - 3 | 839 | 149 | |
AK299081.1 | - | 2 - 3 | 266 | 149 | |
AK315037.1 | - | 4 - 5 | 1098 | 149 | |
AY365125.1 | - | 2 - 3 | 671 | 149 | |
BC036216.1 | - | 4 - 5 | 1127 | 149 | |
BX537641.1 | - | 2 - 3 | 1117 | 149 | |
BX537787.1 | - | 2 - 3 | 986 | 149 | |
BX647096.1 | - | 2 - 3 | 995 | 149 | |
CR936722.1 | - | 2 - 3 | 995 | 149 |