Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAP1GAP
Entrez Gene ID
5909
Gene Name
RAP1 GTPase activating protein
Gene Aliases
RAP1GA1, RAP1GAP1, RAP1GAPII, RAPGAP
Chromosome Location
Chr.1: 21596215 - 21669444 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145657.1 NP_001139129.1 14 - 15 1212 74
NM_001145658.1 NP_001139130.1 13 - 14 1072 74
NM_002885.2 NP_002876.2 13 - 14 1105 74
XM_005245955.3 XP_005246012.2 13 - 14 1117 74
XM_006710804.2 XP_006710867.2 13 - 14 1117 74
XM_006710805.2 XP_006710868.2 13 - 14 1117 74
XM_017001965.1 XP_016857454.1 12 - 13 1119 74
XM_017001966.1 XP_016857455.1 12 - 13 1119 74
XM_017001967.1 XP_016857456.1 13 - 14 1117 74
XM_017001968.1 XP_016857457.1 13 - 14 1152 74
XM_017001969.1 XP_016857458.1 12 - 13 1119 74
XM_017001970.1 XP_016857459.1 12 - 13 1119 74
XM_017001971.1 XP_016857460.1 13 - 14 1071 74
XM_017001972.1 XP_016857461.1 13 - 14 1117 74
XM_017001973.1 XP_016857462.1 12 - 13 1150 74
XM_017001974.1 XP_016857463.1 13 - 14 1117 74
XM_017001975.1 XP_016857464.1 12 - 13 1119 74
XM_017001976.1 XP_016857465.1 13 - 14 1117 74
XM_017001977.1 XP_016857466.1 13 - 14 1117 74
XM_017001978.1 XP_016857467.1 14 - 15 1266 74
XM_017001979.1 XP_016857468.1 15 - 16 1270 74
XM_017001980.1 XP_016857469.1 13 - 14 1071 74
XM_017001981.1 XP_016857470.1 15 - 16 1246 74
XM_017001982.1 XP_016857471.1 14 - 15 1266 74
XM_017001983.1 XP_016857472.1 16 - 17 1478 74
XM_017001984.1 XP_016857473.1 15 - 16 1246 74
XM_017001985.1 XP_016857474.1 13 - 14 1071 74
XM_017001986.1 XP_016857475.1 15 - 16 1283 74
XM_017001987.1 XP_016857476.1 14 - 15 1212 74
XM_017001988.1 XP_016857477.1 14 - 15 1183 74
XM_017001989.1 XP_016857478.1 15 - 16 1283 74
XM_017001990.1 XP_016857479.1 14 - 15 1168 74
XM_017001991.1 XP_016857480.1 14 - 15 1206 74
GenBank mRNA AB007943.3 - 14 - 15 1150 74
BC035030.1 - 10 - 11 861 74
BC054490.1 - 13 - 14 1012 74
BM739021.1 - 6 - 7 496 74
M64788.1 - 13 - 14 1048 74

View Full Product Details