Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAP1GAP
Entrez Gene ID
5909
Gene Name
RAP1 GTPase activating protein
Gene Aliases
RAP1GA1, RAP1GAP1, RAP1GAPII, RAPGAP
Chromosome Location
Chr.1: 21596215 - 21669444 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145657.1 NP_001139129.1 15 - 16 1364 94
NM_001145658.1 NP_001139130.1 14 - 15 1224 94
NM_002885.2 NP_002876.2 14 - 15 1257 94
XM_005245955.3 XP_005246012.2 14 - 15 1269 94
XM_006710804.2 XP_006710867.2 14 - 15 1269 94
XM_006710805.2 XP_006710868.2 14 - 15 1269 94
XM_017001965.1 XP_016857454.1 13 - 14 1271 94
XM_017001966.1 XP_016857455.1 13 - 14 1271 94
XM_017001967.1 XP_016857456.1 14 - 15 1269 94
XM_017001968.1 XP_016857457.1 14 - 15 1304 94
XM_017001969.1 XP_016857458.1 13 - 14 1271 94
XM_017001970.1 XP_016857459.1 13 - 14 1271 94
XM_017001971.1 XP_016857460.1 14 - 15 1223 94
XM_017001972.1 XP_016857461.1 14 - 15 1269 94
XM_017001973.1 XP_016857462.1 13 - 14 1302 94
XM_017001974.1 XP_016857463.1 14 - 15 1269 94
XM_017001975.1 XP_016857464.1 13 - 14 1271 94
XM_017001976.1 XP_016857465.1 14 - 15 1269 94
XM_017001977.1 XP_016857466.1 14 - 15 1269 94
XM_017001978.1 XP_016857467.1 15 - 16 1418 94
XM_017001979.1 XP_016857468.1 16 - 17 1422 94
XM_017001980.1 XP_016857469.1 14 - 15 1223 94
XM_017001981.1 XP_016857470.1 16 - 17 1398 94
XM_017001982.1 XP_016857471.1 15 - 16 1418 94
XM_017001983.1 XP_016857472.1 17 - 18 1630 94
XM_017001984.1 XP_016857473.1 16 - 17 1398 94
XM_017001985.1 XP_016857474.1 14 - 15 1223 94
XM_017001986.1 XP_016857475.1 16 - 17 1435 94
XM_017001987.1 XP_016857476.1 15 - 16 1364 94
XM_017001988.1 XP_016857477.1 15 - 16 1335 94
XM_017001989.1 XP_016857478.1 16 - 17 1435 94
XM_017001990.1 XP_016857479.1 15 - 16 1320 94
XM_017001991.1 XP_016857480.1 15 - 16 1358 94
GenBank mRNA AB007943.3 - 15 - 16 1302 94
BC054490.1 - 14 - 15 1164 94
M64788.1 - 14 - 15 1200 94

View Full Product Details