Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAP1GAP
Entrez Gene ID
5909
Gene Name
RAP1 GTPase activating protein
Gene Aliases
RAP1GA1, RAP1GAP1, RAP1GAPII, RAPGAP
Chromosome Location
Chr.1: 21596215 - 21669444 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145657.1 NP_001139129.1 21 - 22 1985 86
NM_001145658.1 NP_001139130.1 19 - 20 1767 86
NM_002885.2 NP_002876.2 19 - 20 1800 86
XM_005245955.3 XP_005246012.2 20 - 21 1890 86
XM_006710804.2 XP_006710867.2 20 - 21 2067 86
XM_006710805.2 XP_006710868.2 20 - 21 2067 86
XM_017001965.1 XP_016857454.1 19 - 20 2069 86
XM_017001966.1 XP_016857455.1 19 - 20 2069 86
XM_017001968.1 XP_016857457.1 20 - 21 1925 86
XM_017001969.1 XP_016857458.1 19 - 20 1892 86
XM_017001970.1 XP_016857459.1 19 - 20 1892 86
XM_017001971.1 XP_016857460.1 20 - 21 2021 86
XM_017001972.1 XP_016857461.1 19 - 20 1812 86
XM_017001973.1 XP_016857462.1 18 - 19 1845 86
XM_017001975.1 XP_016857464.1 18 - 19 1814 86
XM_017001978.1 XP_016857467.1 21 - 22 2216 86
XM_017001979.1 XP_016857468.1 22 - 23 2220 86
XM_017001980.1 XP_016857469.1 19 - 20 1766 86
XM_017001981.1 XP_016857470.1 22 - 23 2019 86
XM_017001982.1 XP_016857471.1 21 - 22 2039 86
XM_017001983.1 XP_016857472.1 23 - 24 2251 86
XM_017001984.1 XP_016857473.1 22 - 23 2019 86
XM_017001986.1 XP_016857475.1 21 - 22 1978 86
XM_017001987.1 XP_016857476.1 20 - 21 1907 86
XM_017001988.1 XP_016857477.1 20 - 21 1878 86
XM_017001989.1 XP_016857478.1 21 - 22 1978 86
XM_017001990.1 XP_016857479.1 20 - 21 1863 86
GenBank mRNA AB007943.3 - 21 - 22 1923 86
BC035030.1 - 17 - 18 1811 86
BC054490.1 - 19 - 20 1707 86
M64788.1 - 19 - 20 1743 86

View Full Product Details