Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSD3
Entrez Gene ID
23362
Gene Name
pleckstrin and Sec7 domain containing 3
Gene Aliases
EFA6D, EFA6R, HCA67
Chromosome Location
Chr.8: 18527303 - 19086909 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015310.3 NP_056125.3 15 - 16 3025 86
NM_206909.2 NP_996792.1 12 - 13 1429 86
XM_017013258.1 XP_016868747.1 15 - 16 4334 86
XM_017013259.1 XP_016868748.1 15 - 16 4312 86
XM_017013260.1 XP_016868749.1 15 - 16 3085 86
XM_017013261.1 XP_016868750.1 14 - 15 3299 86
XM_017013265.1 XP_016868754.1 12 - 13 1835 86
GenBank mRNA AB023159.2 - 12 - 13 1410 86
AF243495.2 - 13 - 14 1435 86
AF519767.1 - 13 - 14 1383 86
AK091317.1 - 15 - 16 3025 86
AK296546.1 - 7 - 8 1043 86
AK315099.1 - 12 - 13 1457 86
AL832778.1 - 14 - 15 2892 86
BC075044.2 - 12 - 13 1407 86
BC075045.2 - 13 - 14 1410 86
BX647406.1 - 7 - 8 1076 86
GU727648.1 - 15 - 16 2925 86

View Full Product Details