The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145548.1 | NP_001139020.1 | 2 - 3 | 343 | 75 |
NM_017740.2 | NP_060210.2 | 2 - 3 | 343 | 75 | |
XM_011523222.1 | XP_011521524.1 | 2 - 3 | 540 | 75 | |
GenBank mRNA | AK000286.1 | - | 2 - 3 | 343 | 75 |
AK225768.1 | - | 2 - 3 | 295 | 75 | |
BC018772.1 | - | 2 - 3 | 330 | 75 | |
CN427219.1 | - | 2 - 3 | 235 | 75 |