Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNF32
Entrez Gene ID
140545
Gene Name
ring finger protein 32
Gene Aliases
FKSG33, HSD15, LMBR2
Chromosome Location
Chr.7: 156640286 - 156678333 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184996.1 NP_001171925.1 4 - 5 756 83
NM_001184997.1 NP_001171926.1 4 - 5 609 83
NM_001308273.1 NP_001295202.1 4 - 5 707 83
NM_001308274.1 NP_001295203.1 3 - 4 498 83
NM_030936.3 NP_112198.1 4 - 5 651 83
NR_131778.1 4 - 5 605 83
XM_005249522.4 XP_005249579.1 4 - 5 563 83
XM_011515804.2 XP_011514106.1 5 - 6 673 83
XM_011515805.2 XP_011514107.1 5 - 6 830 83
XM_011515806.2 XP_011514108.1 4 - 5 741 83
XM_011515807.2 XP_011514109.1 4 - 5 549 83
XM_011515808.2 XP_011514110.1 4 - 5 661 83
XM_011515809.2 XP_011514111.1 4 - 5 734 83
XM_011515810.2 XP_011514112.1 4 - 5 551 83
XM_011515811.2 XP_011514113.1 4 - 5 546 83
XM_011515812.2 XP_011514114.1 4 - 5 653 83
XM_017011753.1 XP_016867242.1 4 - 5 655 83
XM_017011754.1 XP_016867243.1 4 - 5 655 83
XM_017011755.1 XP_016867244.1 4 - 5 655 83
GenBank mRNA AF325690.1 - 4 - 5 559 83
AF441222.1 - 3 - 4 498 83
AF441224.1 - 4 - 5 605 83
AK307316.1 - 5 - 6 723 83
AK314067.1 - 4 - 5 756 83
AL136874.1 - 4 - 5 539 83
BC015416.1 - 3 - 4 509 83
BT007037.1 - 2 - 3 421 83
CR533513.1 - 2 - 3 421 83

View Full Product Details