Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ADGRL2
- Entrez Gene ID
-
23266
- Gene Name
- adhesion G protein-coupled receptor L2
- Gene Aliases
-
CIRL2, CL2, LEC1, LPHH1, LPHN2
- Chromosome Location
-
Chr.1: 81306155 - 81992738 on Build GRCh38
View Full Product Details