Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHACTR3
Entrez Gene ID
116154
Gene Name
phosphatase and actin regulator 3
Gene Aliases
C20orf101, H17739, PPP1R123, SCAPIN1, SCAPININ
Chromosome Location
Chr.20: 59577497 - 59849829 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199506.1 NP_001186435.1 1 - 2 109 90
GenBank mRNA AJ617581.1 - 1 - 2 109 90
AK316047.1 - 1 - 2 106 90

View Full Product Details