Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DLG1
Entrez Gene ID
1739
Gene Name
discs large MAGUK scaffold protein 1
Gene Aliases
DLGH1, SAP-97, SAP97, dJ1061C18.1.1, hdlg
Chromosome Location
Chr.3: 197042560 - 197299272 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098424.1 NP_001091894.1 8 - 9 883 87
NM_001204386.1 NP_001191315.1 7 - 8 784 87
NM_001204387.1 NP_001191316.1 3 - 4 668 87
NM_001204388.1 NP_001191317.1 3 - 4 668 87
NM_001290983.1 NP_001277912.1 8 - 9 952 87
NM_004087.2 NP_004078.2 8 - 9 883 87
XM_005269289.3 XP_005269346.1 8 - 9 1165 87
XM_011512502.2 XP_011510804.1 8 - 9 1165 87
XM_011512503.1 XP_011510805.1 7 - 8 1063 87
XM_011512505.1 XP_011510807.1 7 - 8 1063 87
XM_011512509.1 XP_011510811.1 7 - 8 1063 87
XM_017005800.1 XP_016861289.1 8 - 9 1300 87
XM_017005801.1 XP_016861290.1 8 - 9 820 87
XM_017005802.1 XP_016861291.1 8 - 9 890 87
XM_017005803.1 XP_016861292.1 8 - 9 811 87
XM_017005804.1 XP_016861293.1 8 - 9 1165 87
XM_017005805.1 XP_016861294.1 8 - 9 982 87
XM_017005806.1 XP_016861295.1 7 - 8 1063 87
XM_017005807.1 XP_016861296.1 7 - 8 1162 87
XM_017005808.1 XP_016861297.1 7 - 8 721 87
XM_017005809.1 XP_016861298.1 7 - 8 823 87
XM_017005810.1 XP_016861299.1 7 - 8 721 87
XM_017005816.1 XP_016861305.1 7 - 8 1162 87
XM_017005817.1 XP_016861306.1 7 - 8 693 87
XM_017005818.1 XP_016861307.1 7 - 8 795 87
XM_017005819.1 XP_016861308.1 7 - 8 712 87
XM_017005821.1 XP_016861310.1 4 - 5 3168 87
XM_017005822.1 XP_016861311.1 4 - 5 3170 87
XM_017005823.1 XP_016861312.1 3 - 4 3065 87
GenBank mRNA AK294855.1 - 3 - 4 668 87
AK310596.1 - 7 - 8 722 87
AK310893.1 - 8 - 9 1167 87
BC015560.2 - 8 - 9 1122 87
BC028486.1 - 8 - 9 1122 87
BC140841.1 - 8 - 9 848 87
BC144651.1 - 7 - 8 749 87
EF553524.1 - 8 - 9 952 87
U13896.1 - 8 - 9 881 87
U13897.1 - 8 - 9 881 87

View Full Product Details