The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323519.1 | NP_001310448.1 | 17 - 18 | 1828 | 66 |
NM_001323521.1 | NP_001310450.1 | 17 - 18 | 1773 | 66 | |
NM_001323522.1 | NP_001310451.1 | 17 - 18 | 1778 | 66 | |
NM_001323523.1 | NP_001310452.1 | 17 - 18 | 2001 | 66 | |
NM_001323524.1 | NP_001310453.1 | 17 - 18 | 1779 | 66 | |
NM_001323526.1 | NP_001310455.1 | 17 - 18 | 1889 | 66 | |
NM_001323532.1 | NP_001310461.1 | 17 - 18 | 1949 | 66 | |
NM_020214.3 | NP_064599.2 | 17 - 18 | 1839 | 66 | |
NR_136594.1 | 18 - 19 | 2070 | 66 | ||
NR_136596.1 | 17 - 18 | 2092 | 66 | ||
NR_136603.1 | 17 - 18 | 1773 | 66 | ||
NR_136604.1 | 18 - 19 | 2010 | 66 | ||
NR_136606.1 | 17 - 18 | 2001 | 66 | ||
NR_136607.1 | 17 - 18 | 1949 | 66 | ||
NR_136608.1 | 17 - 18 | 1839 | 66 | ||
NR_136611.1 | 17 - 18 | 1889 | 66 | ||
XM_017022418.1 | XP_016877907.1 | 16 - 17 | 2429 | 66 | |
GenBank mRNA | AB499727.1 | - | 14 - 15 | 1307 | 66 |
AK024703.1 | - | 5 - 6 | 377 | 66 | |
AL122091.1 | - | 12 - 13 | 1121 | 66 | |
AL390087.1 | - | 15 - 16 | 1499 | 66 | |
BC026955.1 | - | 13 - 14 | 1104 | 66 | |
BC110902.1 | - | 15 - 16 | 1355 | 66 |