The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000930.4 | NP_000921.1 | 3 - 4 | 370 | 83 |
NM_001319189.1 | NP_001306118.1 | 3 - 4 | 370 | 83 | |
GenBank mRNA | A03776.1 | - | 2 - 3 | 147 | 83 |
A07197.1 | - | 2 - 3 | 137 | 83 | |
AK289387.1 | - | 3 - 4 | 322 | 83 | |
AK297738.1 | - | 3 - 4 | 211 | 83 | |
AK297937.1 | - | 3 - 4 | 211 | 83 | |
AK300984.1 | - | 3 - 4 | 211 | 83 | |
AY221101.1 | - | 2 - 3 | 113 | 83 | |
BC007231.1 | - | 3 - 4 | 194 | 83 | |
BC013968.2 | - | 3 - 4 | 196 | 83 | |
BC018636.2 | - | 3 - 4 | 196 | 83 | |
BC095403.1 | - | 3 - 4 | 211 | 83 | |
BT007060.1 | - | 2 - 3 | 113 | 83 | |
DA549782.1 | - | 3 - 4 | 322 | 83 | |
FM956488.1 | - | 2 - 3 | 124 | 83 | |
M15518.1 | - | 3 - 4 | 189 | 83 | |
M18182.1 | - | 2 - 3 | 126 | 83 | |
X07393.1 | - | 3 - 4 | 166 | 83 | |
X13097.1 | - | 3 - 4 | 221 | 83 |