The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001104631.1 | NP_001098101.1 | 14 - 15 | 2190 | 83 |
NM_001165899.1 | NP_001159371.1 | 16 - 17 | 2081 | 83 | |
NM_001197218.1 | NP_001184147.1 | 14 - 15 | 1929 | 83 | |
NM_001197219.1 | NP_001184148.1 | 14 - 15 | 1707 | 83 | |
NM_001197220.1 | NP_001184149.1 | 14 - 15 | 2345 | 83 | |
NM_001197221.1 | NP_001184150.1 | 10 - 11 | 1665 | 83 | |
NM_001197222.1 | NP_001184151.1 | 9 - 10 | 1751 | 83 | |
NM_001197223.1 | NP_001184152.1 | 9 - 10 | 1541 | 83 | |
NM_006203.4 | NP_006194.2 | 14 - 15 | 1733 | 83 | |
XM_005248537.3 | XP_005248594.1 | 14 - 15 | 2887 | 83 | |
XM_005248538.4 | XP_005248595.1 | 15 - 16 | 4788 | 83 | |
XM_011543469.1 | XP_011541771.1 | 17 - 18 | 2170 | 83 | |
XM_011543470.2 | XP_011541772.1 | 17 - 18 | 2094 | 83 | |
XM_011543471.1 | XP_011541773.1 | 16 - 17 | 2047 | 83 | |
XM_011543473.1 | XP_011541775.1 | 16 - 17 | 1995 | 83 | |
XM_011543477.2 | XP_011541779.1 | 16 - 17 | 1997 | 83 | |
XM_017009565.1 | XP_016865054.1 | 17 - 18 | 2207 | 83 | |
XM_017009566.1 | XP_016865055.1 | 17 - 18 | 2097 | 83 | |
XM_017009567.1 | XP_016865056.1 | 15 - 16 | 1835 | 83 | |
XM_017009568.1 | XP_016865057.1 | 17 - 18 | 2081 | 83 | |
XM_017009569.1 | XP_016865058.1 | 16 - 17 | 1960 | 83 | |
XM_017009570.1 | XP_016865059.1 | 13 - 14 | 2780 | 83 | |
XM_017009571.1 | XP_016865060.1 | 16 - 17 | 2003 | 83 | |
GenBank mRNA | AF012073.1 | - | 14 - 15 | 1943 | 83 |
AF012074.1 | - | 10 - 11 | 1679 | 83 | |
AF536975.1 | - | 9 - 10 | 1348 | 83 | |
AF536977.1 | - | 14 - 15 | 1707 | 83 | |
AJ250852.1 | - | 10 - 11 | 1275 | 83 | |
AJ250854.1 | - | 15 - 16 | 1870 | 83 | |
AY245866.1 | - | 16 - 17 | 2046 | 83 | |
AY245867.1 | - | 14 - 15 | 2345 | 83 | |
AY388960.1 | - | 14 - 15 | 1628 | 83 | |
BC036319.1 | - | 9 - 10 | 1543 | 83 | |
L20970.1 | - | 14 - 15 | 1741 | 83 | |
U02882.1 | - | 13 - 14 | 1511 | 83 | |
U50157.1 | - | 9 - 10 | 1349 | 83 | |
U50158.1 | - | 10 - 11 | 1263 | 83 | |
U50159.1 | - | 14 - 15 | 1684 | 83 |