Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VWA5A
Entrez Gene ID
4013
Gene Name
von Willebrand factor A domain containing 5A
Gene Aliases
BCSC-1, BCSC1, LOH11CR2A
Chromosome Location
Chr.11: 124115404 - 124146912 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130142.1 NP_001123614.1 4 - 5 459 81
NM_014622.4 NP_055437.2 3 - 4 344 81
NM_198315.2 NP_938057.1 3 - 4 344 81
XM_011542828.2 XP_011541130.1 4 - 5 570 81
GenBank mRNA AF002672.1 - 2 - 3 257 81
AK291698.1 - 3 - 4 298 81
AK295247.1 - 4 - 5 538 81
AY366503.1 - 3 - 4 344 81
AY366504.1 - 3 - 4 344 81
AY366505.1 - 3 - 4 344 81
AY366506.1 - 4 - 5 459 81
AY366507.1 - 4 - 5 459 81
AY366508.1 - 4 - 5 459 81
BC001234.1 - 3 - 4 321 81
BT006655.1 - 2 - 3 250 81

View Full Product Details