The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289413.1 | NP_001276342.1 | 25 - 26 | 3520 | 67 |
NM_001318833.1 | NP_001305762.1 | 27 - 28 | 3727 | 67 | |
NM_006709.4 | NP_006700.3 | 27 - 28 | 3477 | 67 | |
NM_025256.6 | NP_079532.5 | 26 - 27 | 3375 | 67 | |
XM_005248824.3 | XP_005248881.1 | 26 - 27 | 3618 | 67 | |
XM_006714974.1 | XP_006715037.1 | 26 - 27 | 3643 | 67 | |
XM_006714975.1 | XP_006715038.1 | 25 - 26 | 3541 | 67 | |
XM_006714976.1 | XP_006715039.1 | 27 - 28 | 3481 | 67 | |
XM_017010202.1 | XP_016865691.1 | 26 - 27 | 3379 | 67 | |
GenBank mRNA | AB209433.2 | - | 26 - 27 | 3560 | 67 |
AJ315532.1 | - | 27 - 28 | 3447 | 67 | |
AK092866.1 | - | 10 - 11 | 2392 | 67 | |
AK302904.1 | - | 25 - 26 | 3520 | 67 | |
BC002686.1 | - | 24 - 25 | 2947 | 67 | |
BC009351.1 | - | 27 - 28 | 3444 | 67 | |
BC018718.1 | - | 27 - 28 | 3435 | 67 | |
BC020970.1 | - | 26 - 27 | 3412 | 67 | |
X69838.1 | - | 23 - 24 | 2867 | 67 |