Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EHMT2
Entrez Gene ID
10919
Gene Name
euchromatic histone lysine methyltransferase 2
Gene Aliases
BAT8, C6orf30, G9A, GAT8, KMT1C, NG36
Chromosome Location
Chr.6: 31879759 - 31897707 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289413.1 NP_001276342.1 4 - 5 839 78
NM_001318833.1 NP_001305762.1 5 - 6 923 78
NM_006709.4 NP_006700.3 5 - 6 694 78
NM_025256.6 NP_079532.5 5 - 6 694 78
XM_005248824.3 XP_005248881.1 4 - 5 835 78
XM_006714974.1 XP_006715037.1 4 - 5 839 78
XM_006714975.1 XP_006715038.1 4 - 5 839 78
XM_006714976.1 XP_006715039.1 5 - 6 677 78
XM_017010202.1 XP_016865691.1 5 - 6 677 78
GenBank mRNA AB209433.2 - 4 - 5 756 78
AJ315532.1 - 5 - 6 664 78
AK302904.1 - 4 - 5 839 78
BC002686.1 - 2 - 3 164 78
BC009351.1 - 5 - 6 661 78
BC018718.1 - 5 - 6 652 78
BC020970.1 - 4 - 5 629 78
CN397400.1 - 4 - 5 528 78
DA099526.1 - 2 - 3 304 78
X69838.1 - 1 - 2 84 78

View Full Product Details