The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242464.1 | NP_001229393.1 | 2 - 3 | 188 | 101 |
NM_032192.3 | NP_115568.2 | 2 - 3 | 613 | 101 | |
NM_181505.3 | NP_852606.1 | 2 - 3 | 272 | 101 | |
XM_017025216.1 | XP_016880705.1 | 3 - 4 | 964 | 101 | |
XM_017025217.1 | XP_016880706.1 | 2 - 3 | 84 | 101 | |
GenBank mRNA | AF435972.1 | - | 2 - 3 | 572 | 101 |
AF435973.1 | - | 2 - 3 | 186 | 101 | |
AF435974.1 | - | 2 - 3 | 271 | 101 | |
AF464196.1 | - | 2 - 3 | 611 | 101 | |
AK024593.1 | - | 2 - 3 | 271 | 101 | |
AK123112.1 | - | 2 - 3 | 586 | 101 | |
AK127542.1 | - | 3 - 4 | 477 | 101 | |
AK225058.1 | - | 2 - 3 | 271 | 101 | |
AY070271.1 | - | 2 - 3 | 272 | 101 | |
BC001519.2 | - | 2 - 3 | 249 | 101 | |
BT007076.1 | - | 1 - 2 | 36 | 101 |