Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MED24
Entrez Gene ID
9862
Gene Name
mediator complex subunit 24
Gene Aliases
ARC100, CRSP100, CRSP4, DRIP100, MED5, THRAP4, TRAP100
Chromosome Location
Chr.17: 40019097 - 40054636 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079518.1 NP_001072986.1 10 - 11 1361 91
NM_001267797.1 NP_001254726.1 10 - 11 1339 91
NM_014815.3 NP_055630.2 11 - 12 1378 91
NR_052017.1 10 - 11 1304 91
XM_005257874.2 XP_005257931.1 11 - 12 1283 91
XM_006722204.2 XP_006722267.1 11 - 12 1283 91
XM_006722206.2 XP_006722269.1 11 - 12 1191 91
XM_006722207.2 XP_006722270.1 11 - 12 1159 91
XM_011525529.2 XP_011523831.1 12 - 13 1340 91
XM_011525530.2 XP_011523832.1 12 - 13 1216 91
XM_011525531.2 XP_011523833.1 11 - 12 1159 91
XM_011525532.2 XP_011523834.1 12 - 13 1248 91
XM_011525533.2 XP_011523835.1 12 - 13 1216 91
XM_011525534.2 XP_011523836.1 11 - 12 1159 91
XM_011525535.2 XP_011523837.1 11 - 12 1159 91
XM_011525536.2 XP_011523838.1 12 - 13 1216 91
XM_011525538.2 XP_011523840.1 11 - 12 1474 91
XM_017025458.1 XP_016880947.1 12 - 13 1248 91
XM_017025459.1 XP_016880948.1 11 - 12 1191 91
XM_017025460.1 XP_016880949.1 12 - 13 1248 91
XM_017025461.1 XP_016880950.1 12 - 13 1340 91
XM_017025462.1 XP_016880951.1 12 - 13 1248 91
XM_017025463.1 XP_016880952.1 12 - 13 1216 91
XM_017025464.1 XP_016880953.1 11 - 12 1159 91
XM_017025465.1 XP_016880954.1 11 - 12 1283 91
XM_017025466.1 XP_016880955.1 11 - 12 1191 91
XM_017025467.1 XP_016880956.1 11 - 12 1191 91
XM_017025468.1 XP_016880957.1 11 - 12 1159 91
XM_017025469.1 XP_016880958.1 11 - 12 1474 91
XM_017025470.1 XP_016880959.1 11 - 12 1474 91
XM_017025471.1 XP_016880960.1 11 - 12 1159 91
GenBank mRNA AF055995.1 - 10 - 11 1065 91
AF277379.1 - 10 - 11 1065 91
AK022508.1 - 10 - 11 1130 91
AK291040.1 - 10 - 11 1110 91
AK293309.1 - 9 - 10 900 91
AK299864.1 - 11 - 12 995 91
AK300992.1 - 10 - 11 983 91
BC011375.1 - 10 - 11 1340 91
D50920.1 - 11 - 12 1138 91
EU392550.1 - 4 - 5 243 91

View Full Product Details