Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MED24
Entrez Gene ID
9862
Gene Name
mediator complex subunit 24
Gene Aliases
ARC100, CRSP100, CRSP4, DRIP100, MED5, THRAP4, TRAP100
Chromosome Location
Chr.17: 40019097 - 40054636 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014815.3 NP_055630.2 4 - 5 561 70
XM_005257874.2 XP_005257931.1 4 - 5 466 70
XM_006722204.2 XP_006722267.1 4 - 5 466 70
XM_006722206.2 XP_006722269.1 4 - 5 374 70
XM_006722207.2 XP_006722270.1 4 - 5 342 70
XM_011525529.2 XP_011523831.1 4 - 5 466 70
XM_011525530.2 XP_011523832.1 4 - 5 342 70
XM_011525531.2 XP_011523833.1 4 - 5 342 70
XM_011525532.2 XP_011523834.1 4 - 5 374 70
XM_011525533.2 XP_011523835.1 4 - 5 342 70
XM_011525534.2 XP_011523836.1 3 - 4 285 70
XM_011525535.2 XP_011523837.1 3 - 4 285 70
XM_011525536.2 XP_011523838.1 4 - 5 342 70
XM_011525538.2 XP_011523840.1 3 - 4 285 70
XM_017025458.1 XP_016880947.1 4 - 5 374 70
XM_017025459.1 XP_016880948.1 4 - 5 374 70
XM_017025460.1 XP_016880949.1 4 - 5 374 70
XM_017025461.1 XP_016880950.1 4 - 5 466 70
XM_017025462.1 XP_016880951.1 4 - 5 374 70
XM_017025463.1 XP_016880952.1 4 - 5 342 70
XM_017025464.1 XP_016880953.1 4 - 5 342 70
XM_017025465.1 XP_016880954.1 4 - 5 466 70
XM_017025466.1 XP_016880955.1 4 - 5 374 70
XM_017025467.1 XP_016880956.1 4 - 5 374 70
XM_017025468.1 XP_016880957.1 4 - 5 342 70
XM_017025469.1 XP_016880958.1 3 - 4 285 70
XM_017025470.1 XP_016880959.1 3 - 4 285 70
XM_017025471.1 XP_016880960.1 3 - 4 285 70
GenBank mRNA AF055995.1 - 3 - 4 248 70
AF277379.1 - 3 - 4 248 70
AK300992.1 - 3 - 4 166 70
BC011375.1 - 3 - 4 523 70
D50920.1 - 4 - 5 321 70
EU392534.1 - 2 - 3 115 70
EU392539.1 - 2 - 3 115 70
EU392540.1 - 2 - 3 115 70
EU392543.1 - 2 - 3 115 70
EU392545.1 - 2 - 3 115 70
EU392552.1 - 3 - 4 265 70
EU392553.1 - 2 - 3 115 70

View Full Product Details