Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MED24
- Entrez Gene ID
-
9862
- Gene Name
- mediator complex subunit 24
- Gene Aliases
-
ARC100, CRSP100, CRSP4, DRIP100, MED5, THRAP4, TRAP100
- Chromosome Location
-
Chr.17: 40019097 - 40054636 on Build GRCh38
View Full Product Details