Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KIFC3
Entrez Gene ID
3801
Gene Name
kinesin family member C3
Gene Aliases
-
Chromosome Location
Chr.16: 57758217 - 57863053 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130099.1 NP_001123571.1 6 - 7 1084 97
NM_001130100.1 NP_001123572.1 7 - 8 1140 97
NM_001318710.1 NP_001305639.1 7 - 8 1021 97
NM_001318711.1 NP_001305640.1 7 - 8 1182 97
NM_001318712.1 NP_001305641.1 5 - 6 757 97
NM_001318713.1 NP_001305642.1 5 - 6 1159 97
NM_001318714.1 NP_001305643.1 8 - 9 1529 97
NM_001318715.1 NP_001305644.1 7 - 8 1118 97
NM_005550.3 NP_005541.3 7 - 8 1140 97
NR_134678.1 7 - 8 1324 97
XM_005255937.1 XP_005255994.1 9 - 10 1649 97
XM_005255944.1 XP_005256001.1 7 - 8 1003 97
XM_006721188.1 XP_006721251.1 10 - 11 1727 97
XM_011523075.1 XP_011521377.1 7 - 8 1307 97
XM_011523076.1 XP_011521378.1 7 - 8 1307 97
XM_011523077.1 XP_011521379.1 7 - 8 1106 97
XM_011523078.1 XP_011521380.1 7 - 8 909 97
XM_011523079.1 XP_011521381.1 7 - 8 1244 97
XM_017023219.1 XP_016878708.1 10 - 11 1696 97
XM_017023220.1 XP_016878709.1 10 - 11 1873 97
XM_017023221.1 XP_016878710.1 9 - 10 1649 97
XM_017023222.1 XP_016878711.1 11 - 12 1891 97
XM_017023223.1 XP_016878712.1 7 - 8 1024 97
XM_017023224.1 XP_016878713.1 9 - 10 1558 97
XM_017023225.1 XP_016878714.1 5 - 6 2128 97
XM_017023226.1 XP_016878715.1 8 - 9 1401 97
XM_017023227.1 XP_016878716.1 8 - 9 1401 97
GenBank mRNA AB209290.1 - 7 - 8 1075 97
AF004426.1 - 3 - 4 526 97
AK091891.1 - - 1437 97
AK291737.1 - 6 - 7 979 97
AK296995.1 - 7 - 8 1020 97
AK299431.1 - 7 - 8 1082 97
AK302743.1 - 5 - 6 757 97
AK303021.1 - 5 - 6 1087 97
AK304132.1 - 7 - 8 1128 97
AK307846.1 - 7 - 8 1098 97
BC001211.1 - 7 - 8 1109 97
BC008014.1 - 1 - 2 105 97
BC034234.1 - 8 - 9 1469 97
BC041132.1 - 7 - 8 970 97
BC047051.1 - 7 - 8 1268 97

View Full Product Details