Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ADGRG1
- Entrez Gene ID
-
9289
- Gene Name
- adhesion G protein-coupled receptor G1
- Gene Aliases
-
BFPP, BPPR, GPR56, TM7LN4, TM7XN1
- Chromosome Location
-
Chr.16: 57619535 - 57665039 on Build GRCh38
View Full Product Details