Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KNTC1
Entrez Gene ID
9735
Gene Name
kinetochore associated 1
Gene Aliases
ROD
Chromosome Location
Chr.12: 122527114 - 122626409 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014708.4 NP_055523.1 50 - 51 5547 142
XM_006719706.2 XP_006719769.1 50 - 51 5529 142
XM_011539027.2 XP_011537329.1 50 - 51 5529 142
XM_017020254.1 XP_016875743.1 49 - 50 5439 142
XM_017020255.1 XP_016875744.1 49 - 50 5493 142
XM_017020256.1 XP_016875745.1 31 - 32 3803 142
GenBank mRNA AK093470.1 - 5 - 6 903 142
AK293403.1 - 7 - 8 1035 142
BC150278.1 - 50 - 51 5547 142
D79988.1 - 50 - 51 5547 142

View Full Product Details