The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014708.4 | NP_055523.1 | 56 - 57 | 6132 | 107 |
XM_006719706.2 | XP_006719769.1 | 56 - 57 | 6114 | 107 | |
XM_011539027.2 | XP_011537329.1 | 56 - 57 | 6036 | 107 | |
XM_017020254.1 | XP_016875743.1 | 55 - 56 | 6024 | 107 | |
XM_017020255.1 | XP_016875744.1 | 55 - 56 | 6078 | 107 | |
XM_017020256.1 | XP_016875745.1 | 37 - 38 | 4388 | 107 | |
GenBank mRNA | AK293403.1 | - | 13 - 14 | 1620 | 107 |
AK303337.1 | - | 31 - 32 | 2889 | 107 | |
BC150278.1 | - | 56 - 57 | 6132 | 107 | |
D79988.1 | - | 56 - 57 | 6132 | 107 |