The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031719.2 | NP_001026889.1 | 7 - 8 | 676 | 93 |
XM_011535235.2 | XP_011533537.1 | 9 - 10 | 881 | 93 | |
XM_011535236.2 | XP_011533538.1 | 8 - 9 | 812 | 93 | |
XM_011535237.1 | XP_011533539.1 | 8 - 9 | 760 | 93 | |
XM_011535238.1 | XP_011533540.1 | 7 - 8 | 691 | 93 | |
XM_017020753.1 | XP_016876242.1 | 6 - 7 | 620 | 93 | |
GenBank mRNA | BC009825.2 | - | 7 - 8 | 612 | 93 |