The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017952.5 | NP_060422.4 | 22 - 23 | 1884 | 138 |
GenBank mRNA | AK000765.1 | - | 12 - 13 | 1055 | 138 |
AK315676.1 | - | 16 - 17 | 1338 | 138 | |
AL832520.1 | - | 11 - 12 | 888 | 138 | |
AY277598.1 | - | 10 - 11 | 852 | 138 | |
BC001758.1 | - | 11 - 12 | 958 | 138 | |
BC011832.2 | - | 22 - 23 | 1819 | 138 |