The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083592.1 | NP_001077061.1 | 3 - 4 | 849 | 68 |
NM_005012.3 | NP_005003.2 | 3 - 4 | 849 | 68 | |
XM_011541526.1 | XP_011539828.1 | 3 - 4 | 448 | 68 | |
XM_017001376.1 | XP_016856865.1 | 2 - 3 | 749 | 68 | |
XM_017001377.1 | XP_016856866.1 | 5 - 6 | 3392 | 68 | |
GenBank mRNA | AK308088.1 | - | 3 - 4 | 855 | 68 |
BC080541.1 | - | 3 - 4 | 849 | 68 | |
BC128386.1 | - | 4 - 5 | 459 | 68 | |
M97675.1 | - | 3 - 4 | 825 | 68 |