Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRAFD1
Entrez Gene ID
10906
Gene Name
TRAF-type zinc finger domain containing 1
Gene Aliases
FLN29
Chromosome Location
Chr.12: 112125545 - 112153604 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143906.1 NP_001137378.1 3 - 4 796 81
NM_006700.2 NP_006691.1 3 - 4 265 81
GenBank mRNA AB007447.1 - 3 - 4 233 81
AK122620.1 - 3 - 4 781 81
AK291331.1 - 3 - 4 250 81
AK295474.1 - 3 - 4 250 81
AK309916.1 - 3 - 4 272 81
AK312601.1 - 3 - 4 265 81
BC003553.2 - 2 - 3 191 81

View Full Product Details