Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRAFD1
Entrez Gene ID
10906
Gene Name
TRAF-type zinc finger domain containing 1
Gene Aliases
FLN29
Chromosome Location
Chr.12: 112125545 - 112153604 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143906.1 NP_001137378.1 7 - 8 1547 64
NM_006700.2 NP_006691.1 7 - 8 1016 64
GenBank mRNA AB007447.1 - 7 - 8 984 64
AK122620.1 - 7 - 8 1531 64
AK291331.1 - 7 - 8 1001 64
AK295474.1 - 7 - 8 757 64
AK312601.1 - 7 - 8 1016 64
BC003553.2 - 6 - 7 942 64

View Full Product Details