The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145800.3 | NP_665799.1 | 10 - 11 | 1588 | 69 |
XM_011531317.2 | XP_011529619.1 | 10 - 11 | 1413 | 69 | |
GenBank mRNA | AF403060.1 | - | 10 - 11 | 1588 | 69 |
AK026589.1 | - | 10 - 11 | 1381 | 69 |