The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006558.2 | NP_006549.1 | 8 - 9 | 1347 | 72 |
XM_005250757.2 | XP_005250814.1 | 8 - 9 | 1039 | 72 | |
XM_011516796.2 | XP_011515098.1 | 8 - 9 | 1189 | 72 | |
XM_011516797.2 | XP_011515099.1 | 8 - 9 | 1224 | 72 | |
GenBank mRNA | AF051321.1 | - | 8 - 9 | 1339 | 72 |
AF051322.1 | - | 7 - 8 | 841 | 72 | |
AF069681.1 | - | 8 - 9 | 1295 | 72 | |
BC032606.2 | - | 8 - 9 | 1356 | 72 | |
BC068536.1 | - | 9 - 10 | 1148 | 72 |