The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003215.2 | NP_003206.2 | 12 - 13 | 1169 | 99 |
XM_011513737.2 | XP_011512039.1 | 12 - 13 | 1150 | 99 | |
XM_011513741.1 | XP_011512043.1 | 11 - 12 | 1123 | 99 | |
XM_017008569.1 | XP_016864058.1 | 11 - 12 | 2375 | 99 | |
GenBank mRNA | BC101711.1 | - | 12 - 13 | 1177 | 99 |
BC101713.1 | - | 12 - 13 | 1177 | 99 | |
BC143487.1 | - | 12 - 13 | 1177 | 99 | |
D29767.1 | - | 12 - 13 | 1195 | 99 |