Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TEC
Entrez Gene ID
7006
Gene Name
tec protein tyrosine kinase
Gene Aliases
PSCTK4
Chromosome Location
Chr.4: 48135783 - 48269864 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003215.2 NP_003206.2 8 - 9 835 138
XM_011513737.2 XP_011512039.1 8 - 9 816 138
XM_011513741.1 XP_011512043.1 7 - 8 789 138
XM_017008569.1 XP_016864058.1 7 - 8 2041 138
GenBank mRNA BC101711.1 - 8 - 9 843 138
BC101713.1 - 8 - 9 843 138
BC143487.1 - 8 - 9 843 138

View Full Product Details