The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322902.1 | NP_001309831.1 | 6 - 7 | 686 | 58 |
NM_001322903.1 | NP_001309832.1 | 7 - 8 | 782 | 58 | |
NM_001322904.1 | NP_001309833.1 | 7 - 8 | 752 | 58 | |
NM_001322905.1 | NP_001309834.1 | 7 - 8 | 715 | 58 | |
NM_001322906.1 | NP_001309835.1 | 7 - 8 | 709 | 58 | |
NM_001322907.1 | NP_001309836.1 | 8 - 9 | 910 | 58 | |
NM_001322908.1 | NP_001309837.1 | 8 - 9 | 833 | 58 | |
NM_001322909.1 | NP_001309838.1 | 7 - 8 | 700 | 58 | |
NM_001322910.1 | NP_001309839.1 | 7 - 8 | 807 | 58 | |
NM_003081.4 | NP_003072.2 | 6 - 7 | 655 | 58 | |
NM_130811.3 | NP_570824.1 | 6 - 7 | 655 | 58 | |
XM_005260808.4 | XP_005260865.1 | 6 - 7 | 733 | 58 | |
XM_017028021.1 | XP_016883510.1 | 7 - 8 | 585 | 58 | |
XM_017028022.1 | XP_016883511.1 | 7 - 8 | 677 | 58 | |
GenBank mRNA | AK026732.1 | - | 1 - 2 | 826 | 58 |
AK090857.1 | - | 1 - 2 | 1623 | 58 | |
AK094560.1 | - | 7 - 8 | 741 | 58 | |
AK098767.1 | - | 6 - 7 | 525 | 58 | |
AK223617.1 | - | 6 - 7 | 617 | 58 | |
AK289647.1 | - | 8 - 9 | 816 | 58 | |
AK314359.1 | - | 6 - 7 | 566 | 58 | |
BC010647.1 | - | 6 - 7 | 544 | 58 | |
BI604120.1 | - | 6 - 7 | 634 | 58 | |
BT019684.1 | - | 5 - 6 | 412 | 58 | |
BX647443.1 | - | 6 - 7 | 527 | 58 | |
D21267.1 | - | 6 - 7 | 617 | 58 | |
L19760.1 | - | 6 - 7 | 500 | 58 | |
L19761.1 | - | 6 - 7 | 500 | 58 |