The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005440.4 | NP_005431.1 | 2 - 3 | 293 | 64 |
XM_011525316.1 | XP_011523618.1 | 2 - 3 | 293 | 64 | |
XM_011525317.2 | XP_011523619.1 | 2 - 3 | 133 | 64 | |
GenBank mRNA | AB209778.1 | - | 2 - 3 | 144 | 64 |
AF498968.1 | - | 2 - 3 | 186 | 64 | |
AK290199.1 | - | 2 - 3 | 298 | 64 | |
BC018096.1 | - | 2 - 3 | 182 | 64 | |
BC094842.1 | - | 2 - 3 | 286 | 64 | |
BC104986.1 | - | 2 - 3 | 265 | 64 | |
BC104990.1 | - | 2 - 3 | 265 | 64 | |
BT019394.1 | - | 2 - 3 | 186 | 64 | |
X95456.1 | - | 2 - 3 | 186 | 64 |