The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303037.1 | NP_001289966.1 | 10 - 11 | 1572 | 89 |
NM_016447.3 | NP_057531.2 | 11 - 12 | 1741 | 89 | |
XM_005249775.4 | XP_005249832.1 | 9 - 10 | 1353 | 89 | |
XM_006715738.3 | XP_006715801.1 | 10 - 11 | 1872 | 89 | |
XM_006715739.3 | XP_006715802.1 | 10 - 11 | 1463 | 89 | |
XM_006715740.3 | XP_006715803.1 | 10 - 11 | 1460 | 89 | |
XM_011515425.2 | XP_011513727.1 | 10 - 11 | 1568 | 89 | |
XM_017012315.1 | XP_016867804.1 | 10 - 11 | 1496 | 89 | |
XM_017012316.1 | XP_016867805.1 | 10 - 11 | 1617 | 89 | |
XM_017012317.1 | XP_016867806.1 | 10 - 11 | 1497 | 89 | |
GenBank mRNA | AF162130.2 | - | 10 - 11 | 1513 | 89 |
AL136836.1 | - | 11 - 12 | 1621 | 89 | |
BC023638.2 | - | 10 - 11 | 1536 | 89 |