Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRPPRC
Entrez Gene ID
10128
Gene Name
leucine rich pentatricopeptide repeat containing
Gene Aliases
CLONE-23970, GP130, LRP130, LSFC
Chromosome Location
Chr.2: 43886224 - 43996005 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133259.3 NP_573566.2 23 - 24 2564 60
XM_006711915.2 XP_006711978.1 23 - 24 2573 60
XM_006711916.3 XP_006711979.1 23 - 24 2548 60
XM_011532473.1 XP_011530775.1 23 - 24 2548 60
XM_017003117.1 XP_016858606.1 23 - 24 2546 60
GenBank mRNA AK125781.1 - 23 - 24 2535 60
AK290016.1 - 23 - 24 2535 60
AK299869.1 - 22 - 23 2336 60
AY289212.1 - 23 - 24 2564 60
BC010282.1 - 3 - 4 431 60
BC050311.1 - 23 - 24 2515 60
BC130285.1 - 23 - 24 2560 60
M92439.1 - 22 - 23 2188 60

View Full Product Details