The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133259.3 | NP_573566.2 | 27 - 28 | 2957 | 107 |
XM_006711915.2 | XP_006711978.1 | 27 - 28 | 2966 | 107 | |
XM_006711916.3 | XP_006711979.1 | 27 - 28 | 2941 | 107 | |
XM_011532473.1 | XP_011530775.1 | 27 - 28 | 2941 | 107 | |
XM_017003117.1 | XP_016858606.1 | 27 - 28 | 2939 | 107 | |
GenBank mRNA | AK125781.1 | - | 27 - 28 | 2928 | 107 |
AK290016.1 | - | 27 - 28 | 2928 | 107 | |
AK299869.1 | - | 26 - 27 | 2729 | 107 | |
AY289212.1 | - | 27 - 28 | 2957 | 107 | |
BC010282.1 | - | 7 - 8 | 824 | 107 | |
BC050311.1 | - | 27 - 28 | 2908 | 107 | |
BC130285.1 | - | 27 - 28 | 2953 | 107 | |
M92439.1 | - | 26 - 27 | 2581 | 107 |