Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRPPRC
Entrez Gene ID
10128
Gene Name
leucine rich pentatricopeptide repeat containing
Gene Aliases
CLONE-23970, GP130, LRP130, LSFC
Chromosome Location
Chr.2: 43886224 - 43996005 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133259.3 NP_573566.2 5 - 6 705 116
XM_006711915.2 XP_006711978.1 5 - 6 714 116
XM_006711916.3 XP_006711979.1 5 - 6 689 116
XM_011532473.1 XP_011530775.1 5 - 6 689 116
XM_017003117.1 XP_016858606.1 5 - 6 687 116
GenBank mRNA AK125781.1 - 5 - 6 676 116
AK290016.1 - 5 - 6 676 116
AK299869.1 - 4 - 5 477 116
AK308348.1 - 5 - 6 701 116
AY289212.1 - 5 - 6 705 116
BC026034.1 - 5 - 6 678 116
BC050311.1 - 5 - 6 656 116
BC130285.1 - 5 - 6 701 116
M92439.1 - 4 - 5 329 116

View Full Product Details