Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCAMP1
Entrez Gene ID
9522
Gene Name
secretory carrier membrane protein 1
Gene Aliases
SCAMP, SCAMP37
Chromosome Location
Chr.5: 78360503 - 78480739 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290229.1 NP_001277158.1 5 - 6 722 121
NM_004866.5 NP_004857.4 6 - 7 800 121
GenBank mRNA AF005037.1 - 6 - 7 674 121
AF038966.1 - 6 - 7 721 121
AK225172.1 - 6 - 7 689 121
AK290225.1 - 6 - 7 650 121
AK298146.1 - 4 - 5 433 121
AK304452.1 - 5 - 6 614 121
BC015065.1 - 6 - 7 693 121
BC034048.1 - 6 - 7 654 121
CR542286.1 - 6 - 7 631 121

View Full Product Details