The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006184.5 | NP_006175.2 | 10 - 11 | 1333 | 96 |
XM_017026845.1 | XP_016882334.1 | 10 - 11 | 1300 | 96 | |
GenBank mRNA | AK025458.1 | - | 7 - 8 | 762 | 96 |
AK097790.1 | - | 1 - 2 | 285 | 96 | |
AK222805.1 | - | 10 - 11 | 1040 | 96 | |
AK292066.1 | - | 10 - 11 | 1333 | 96 | |
AK303126.1 | - | 10 - 11 | 1009 | 96 | |
AK315422.1 | - | 10 - 11 | 1071 | 96 | |
BC002356.2 | - | 9 - 10 | 1016 | 96 | |
BC015787.1 | - | 4 - 5 | 298 | 96 | |
BT009828.1 | - | 9 - 10 | 999 | 96 | |
BX647291.1 | - | 7 - 8 | 615 | 96 | |
M96824.1 | - | 10 - 11 | 1035 | 96 | |
U92978.1 | - | 2 - 3 | 153 | 96 |